Merck
  • Synthesis and study of 2-amino-7-bromofluorenes modified with nitroxides and their precursors as dual anti-amyloid and antioxidant active compounds.
Synthesis and study of 2-amino-7-bromofluorenes modified with nitroxides and their precursors as dual anti-amyloid and antioxidant active compounds.
European journal of medicinal chemistry (2011-02-22)
Tamás Kálai, Jitka Petrlova, Mária Balog, Hnin Hnin Aung, John C Voss, Kálmán Hideg
ABSTRACT
A series of 2-aminofluorenes N-alkylated with nitroxides or their precursors were synthesized. The new compounds were tested on hydroxyl radical and peroxyl radical scavenging ability and inflammatory assay on the endothelial brain cells. In agreement with ROS scavenging ability the same compound 7-bromo-N -[(1-Oxyl-2,2,6,6-tetramethyl-1,2,3,6-tetrahydropyridine-4yl)methyl]-9H-fluoren-2-amine (3b) and its hydroxylamine salt (3b/OH/HCl) showed the anti-inflammatory property on the endothelial brain cells.
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Sigma-Aldrich
4-Hydroxy-TEMPO, 95%