Merck
  • Detection of feline herpes virus 1 via polymerase chain reaction and immunohistochemistry in cats with ulcerative facial dermatitis, eosinophilic granuloma complex reaction patterns and mosquito bite hypersensitivity.
Detection of feline herpes virus 1 via polymerase chain reaction and immunohistochemistry in cats with ulcerative facial dermatitis, eosinophilic granuloma complex reaction patterns and mosquito bite hypersensitivity.
Veterinary dermatology (2011-06-10)
Paola Persico, Paola Roccabianca, Antonio Corona, Antonella Vercelli, Luisa Cornegliani
ABSTRACT
Ulcerative dermatitis caused by feline herpes virus 1 (FHV-1) is an uncommon disease characterized by cutaneous ulcers secondary to epidermal, adnexal and dermal necrosis. Differential diagnoses for FHV-1 lesions include, but are not limited to, mosquito bite hypersensitivity and eosinophilic granuloma complex. Histopathological diagnosis of FHV-1 dermatitis is based on the detection of the intranuclear inclusion bodies. In cases where intranuclear inclusions are missing but clinical and histological findings are compatible with FHV-1 dermatitis, immunohistochemistry (IHC) and PCRs have been used. In this retrospective study, we evaluated the presence of FHV-1 by IHC and PCR in skin biopsies and compared the results of the two tests. Sixty-four skin biopsy specimens from cats with compatible lesions were reviewed and tested via PCR and IHC for evidence of FHV-1. Polymerase chain reaction was positive in 12 of 64 biopsies; PCR and IHC were positive only in two of 64 biopsies, and these cases were considered true positive cases. The higher number of PCR-positive cases was possibly attributed to amplification of viral DNA from a live attenuated vaccination, but a previous FHV-1 infection with subsequent amplification of latently inserted FHV-1 could not be excluded. If clinical signs and histopathology suggest FHV-1 infection in the absence of typical inclusion bodies, IHC is the preferred diagnostic test; PCR may be useful for initial screening, but due to false positives is not sufficient for a definitive diagnosis.
Sigma-Aldrich
REDTaq® Genomic DNA Polymerase, without MgCl2
Pricing and availability is not currently available.