Merck
  • Protein transport in human cells mediated by covalently and noncovalently conjugated arginine-rich intracellular delivery peptides.
Protein transport in human cells mediated by covalently and noncovalently conjugated arginine-rich intracellular delivery peptides.
Peptides (2009-06-16)
Jia-Wei Hu, Betty Revon Liu, Chih-Yuan Wu, Shu-Wan Lu, Han-Jung Lee
ABSTRACT
Generally, biomacromolecules, such as DNA, RNA, and proteins, cannot freely permeate into cells from outside the membrane. Protein transduction domains (PTDs) are peptides containing a large number of basic amino acids that can deliver macromolecules into living cells. Arginine-rich intracellular delivery (AID) peptides are more effective than other PTD peptides at carrying large molecules across cellular membranes. In the present study, we demonstrated that AID peptides are able to deliver cargo proteins into living cells in both covalent and noncovalent protein transductions (CNPT) synchronously. Human A549 cells were treated with a fluorescent protein (FP) that was noncovalently premixed with another AID-conjugated FP, which emitted a different color. After the delivery of carrier AID-FP and cargo FP into cells, the emission and merge of fluorescence were observed and recorded with a confocal microscope, while the internalization efficiency was quantitatively analyzed with a flow cytometer. The optimal molecular ratio between carrier AID-FP and cargo FP for CNPT is about 1:1/3. Fluorescence resonance energy transfer (FRET) assay further confirmed AID-conjugates can physically interact with its cargo FPs in CNPT in cells. Potential uptake mechanisms of CNPT may involve a combination of multiple internalization pathways. After delivery, intracellular distributions of AID-conjugates and FPs may possibly colocalize with lysosomes. These results will facilitate the understanding of multiple mechanisms of PTDs, and provide a powerful tool for simultaneously delivering several proteins or compounds in protein internalization.
Sigma-Aldrich
Sodium chlorate, ACS reagent, ≥99.0%