Merck
  • Asymmetric Michael addition of substituted rhodanines to α,β-unsaturated ketones catalyzed by bulky primary amines.
Asymmetric Michael addition of substituted rhodanines to α,β-unsaturated ketones catalyzed by bulky primary amines.
Organic letters (2012-04-10)
Feng Yu, Haoxiang Hu, Xiaodong Gu, Jinxing Ye
ABSTRACT
A bulky group was introduced by design into a diamine catalyst, and a series of robust and tunable bulky chiral primary amine catalysts were developed and successfully applied in the direct conjugate addition of substituted rhodanines to α,β-unsaturated ketones. High yields (up to 99%) and excellent diastereoselectivities (up to 99:1 dr) and enantioselectivities (up to 98% ee) were observed.
Sigma-Aldrich
Rhodanine, 97%