Merck
  • 1H and 13C NMR assignments for the cyanine dyes SYBR Safe and thiazole orange.
1H and 13C NMR assignments for the cyanine dyes SYBR Safe and thiazole orange.
The Journal of organic chemistry (2012-11-10)
William E Evenson, Lauren M Boden, Katy A Muzikar, Daniel J O'Leary
ABSTRACT
Analysis of (1)H and (13)C NMR and mass spectral data for the fluorescent nucleic acid stain SYBR Safe indicates that it contains a cyanine-based cationic core structure identical to thiazole orange. The difference between these two compounds is the type of N-substitution on the quinolinium ring system (SYBR Safe, n-Pr; thiazole orange, Me). The (1)H and (13)C NMR resonances for both compounds were assigned on the basis of one- and two-dimensional (COSY, ROESY, HSQC, and HMBC) experiments. The preferred conformation of these compounds was computed by ab initio methods and found to be consistent with the NMR data.
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %