Merck
  • Multivalent glycoclusters constructed by chiral self-assembly of mannose functionalized perylene bisimide.
Multivalent glycoclusters constructed by chiral self-assembly of mannose functionalized perylene bisimide.
Organic & biomolecular chemistry (2013-01-08)
Ke-Rang Wang, Hong-Wei An, Yue-Qing Wang, Jin-Chao Zhang, Xiao-Liu Li
ABSTRACT
Water-soluble perylene bisimide derivative 7 modified with six mannoses was synthesized and its self-assembled properties were studied by UV-Vis and CD spectroscopy, which revealed an interesting self-assembly with a solvent-tuning chiral conformation in H(2)O-DMSO solution. As H(2)O was added to the DMSO solution until a 60% (or 70%) v/v proportion was achieved, the self-assembly of the mannose functionalized compound 7 exhibited a left-handed helical conformation. More interestingly, when the volume of H(2)O constituted beyond 85% of the solution, the conformation of the self-assembly turned out to be a right-handed helical conformation. Furthermore, the binding interactions between the self-assembly of compound 7 and Con A were investigated by turbidity assay, CD spectra, TEM and SEM images, and ELLA experiment, which indicated that the self-assembly of compound 7 as multivalent glycoclusters exhibited specific binding to Con A with an IC(50) value of 24 μM (144 μM, valency corrected), 10 times stronger than the reference compound (α-MMP).
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture