Merck
Genetic toxicological testing of some dyes by the micronucleus test.
Mutation research (1986-04-01)
R N Misra, B Misra
ABSTRACT
Three widely used dyes, acridine orange, blue VRS and fast green FCF were administered to male mice in order to study the induction of gross chromosomal anomalies using the micronucleus test. All 3 compounds were shown to be clastogenic.
Product Description
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
Fast Green FCF, certified by the BSC
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %