Merck
  • Aberrant cyclization affords a C-6 modified cyclic adenosine 5'-diphosphoribose analogue with biological activity in Jurkat T cells.
Aberrant cyclization affords a C-6 modified cyclic adenosine 5'-diphosphoribose analogue with biological activity in Jurkat T cells.
Journal of medicinal chemistry (2012-01-18)
Christelle Moreau, Tanja Kirchberger, Bo Zhang, Mark P Thomas, Karin Weber, Andreas H Guse, Barry V L Potter
ABSTRACT
Two nicotinamide adenine dinucleotide (NAD(+)) analogues modified at the 6 position of the purine ring were synthesized, and their substrate properties toward Aplysia californica ADP-ribosyl cyclase were investigated. 6-N-Methyl NAD(+) (6-N-methyl nicotinamide adenosine 5'-dinucleotide 10) hydrolyzes to give the linear 6-N-methyl ADPR (adenosine 5'-diphosphoribose, 11), whereas 6-thio NHD(+) (nicotinamide 6-mercaptopurine 5'-dinucleotide, 17) generates a cyclic dinucleotide. Surprisingly, NMR correlation spectra confirm this compound to be the N1 cyclic product 6-thio N1-cIDPR (6-thio cyclic inosine 5'-diphosphoribose, 3), although the corresponding 6-oxo analogue is well-known to cyclize at N7. In Jurkat T cells, unlike the parent cyclic inosine 5'-diphosphoribose N1-cIDPR 2, 6-thio N1-cIDPR antagonizes both cADPR- and N1-cIDPR-induced Ca(2+) release but possesses weak agonist activity at higher concentration. 3 is thus identified as the first C-6 modified cADPR (cyclic adenosine 5'-diphosphoribose) analogue antagonist; it represents the first example of a fluorescent N1-cyclized cADPR analogue and is a new pharmacological tool for intervention in the cADPR pathway of cellular signaling.
Sigma-Aldrich
Cyclic adenosine diphosphate-ribose, ≥90% (HPLC), lyophilized powder