Merck
  • Simple O-acylated derivatives of lysergol and dihydrolysergol-I: synthesis and interaction with 5-HT2A, 5-HT2C and 5-HT1B receptors, and alpha1 adrenergic receptors.
Simple O-acylated derivatives of lysergol and dihydrolysergol-I: synthesis and interaction with 5-HT2A, 5-HT2C and 5-HT1B receptors, and alpha1 adrenergic receptors.
The Journal of pharmacy and pharmacology (1999-05-27)
H H Pertz, A M Brown, T L Gager, A J Kaumann
ABSTRACT
A series of simple O-acylated derivatives of the naturally occurring clavine alkaloids lysergol and dihydrolysergol-I were synthesized and tested in-vitro for their ability to interact with 5-HT2A receptors in rat tail artery, 5-HT2C receptors in piglet choroid plexus, 5-HT1B receptors in guinea-pig iliac artery and alpha1-adrenergic receptors in rat aorta. In contrast to the classical ergoline 5-HT2A receptor antagonists methysergide and LY53857, the compounds produced competitive antagonism of the 5-HT response in rat tail artery. Affinities of ergolines 3-14 were higher (pA2 values of 7.33-8.40) than those of the parent alcohols lysergol (1) and dihydrolysergol-I (2), respectively. The introduction of an isopropyl substituent at the N(1) position of the compounds failed to enhance 5-HT2A receptor affinity. Compounds 3-14 exhibited lower affinities for alpha1-adrenergic receptors than for 5-HT2A receptors. In particular, those lysergol derivatives that had an isopropyl substituent at the N(1) position were highly specific 5-HT2A receptor antagonists (ratio 5-HT2A/alpha1 = 302-3548). Selected derivatives of lysergol (3-5, 9-11) which were assayed for radioligand binding at 5-HT2C receptors in piglet choroid plexus had affinities that were similar to those found in rat tail artery. Additionally, lysergol and its N(1)-unsubstituted derivatives were found to be partial agonists (alpha of 0.2-0.4) for 5-HT2C receptor-mediated inositol phosphate accumulation in piglet choroid plexus. On the other hand, analogues with an isopropyl substituent at N(1) showed no measurable agonist activity. The observation that N(1)-unsubstituted derivatives of lysergol possessed agonist properties at 5-HT2C receptors whereas their agonist activity at 5-HT2A receptors was marginal (alpha of 0.05 for compound 3 at 1 microM) or not measurable, suggests that these compounds have different abilities to cause conformational change at the two receptor types. Selected derivatives of lysergol (3-5, 9-11) which were examined as ligands for 5-HT1B receptors in guinea-pig iliac artery caused insurmountable blockade of the contractile effect of 5-HT. N(1)-isopropyl derivatives had 30-50-fold lower affinities for 5-HT1B receptors of this tissue than their N(1)-unsubstituted analogues. It is concluded that O-acylated derivatives of the clavine alkaloids lysergol and dihydrolysergol-I mimic therapeutically relevant ergolines due to the complexity of their pharmacological profile as partial agonists and antagonists at 5-HT2A, 5-HT2C and 5-HT1B receptors, and at alpha1-adrenergic receptors.