Merck
Rhodium(II)-catalyzed stereoselective synthesis of allylsilanes.
Organic letters (2013-11-29)
David M Guptill, Carolyn M Cohen, Huw M L Davies
ABSTRACT
The rhodium-catalyzed decomposition of 2-(triisopropylsilyl)ethyl aryl- and vinyldiazoacetates results in the stereoselective formation of Z-allylsilanes. The transformation is considered to proceed by silyl-directed intramolecular C-H functionalization to form a β-lactone intermediate followed by a silyl-activated extrusion of carbon dioxide.
Sigma-Aldrich
Rh2(R-BTPCP)4
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis