Merck
  • A click chemistry approach to tetrazoles by Huisgen 1,3-dipolar cycloaddition: synthesis of 5-acyltetrazoles from azides and acyl cyanides.
A click chemistry approach to tetrazoles by Huisgen 1,3-dipolar cycloaddition: synthesis of 5-acyltetrazoles from azides and acyl cyanides.
Angewandte Chemie (International ed. in English) (2002-06-17)
Zachary P Demko, K Barry Sharpless
Sigma-Aldrich
p-Toluenesulfonyl cyanide, technical grade, 95%