Merck
  • Anti-diabetic activity of swertiamarin is due to an active metabolite, gentianine, that upregulates PPAR-γ gene expression in 3T3-L1 cells.
Anti-diabetic activity of swertiamarin is due to an active metabolite, gentianine, that upregulates PPAR-γ gene expression in 3T3-L1 cells.
Phytotherapy research : PTR (2012-06-22)
Hitesh Vaidya, Ramesh K Goyal, Sukhinder Kaur Cheema
ABSTRACT
We have previously shown the anti-diabetic effects of swertiamarin; however, pharmacokinetic analysis showed that swertiamarin had a plasma half-life of 1.3 h. Gentianine is an active metabolite of swertiamarin that possesses a pharmacophoric moiety. The aim of this study was to explore the possibility whether the anti-diabetic effect of swertiamarin is due to gentianine. Swertiamarin treatment had no significant effect on adipogenesis, or the mRNA expression of PPAR-γ and GLUT-4; however, there was a significant increase in the mRNA expression of adiponectin. On the other hand, treatment with gentianine significantly increased adipogenesis, which was associated with a significant increase in the mRNA expression of PPAR-γ, GLUT-4 and adiponectin. These findings suggest, for the first time, that the anti-diabetic effect of swertiamarin is due to gentianine, an active metabolite of swertiamarin.
Sigma-Aldrich
Swertiamarin, ≥95% (LC/MS-ELSD)
Supelco
Swertiamarin, analytical standard