Merck
  • Development of antibodies for determination of alkylresorcinol metabolites in human urine and elucidation of ELISA cross-reactivity.
Development of antibodies for determination of alkylresorcinol metabolites in human urine and elucidation of ELISA cross-reactivity.
Journal of immunological methods (2014-07-30)
R Wierzbicka, L Eyer, R Landberg, A Kamal-Eldin, M Franek
ABSTRACT
Alkylresorcinols (ARs) are amphiphilic phenolic lipids and their two main metabolites, 3-(3,5-dihydroxyphenyl)-propanoic acid (DHPPA) and 3,5-dihydroxybenzoic acid (DHBA), can be used as biomarkers of whole grain wheat and rye intake. The aim of this study was to develop antibodies against DHBA and DHPPA for use in ELISA analysis. Good calibration curves were obtained for ELISA using alkaline phosphatase (AP) conjugates. The highest sensitivity for DHPPA was found using a reagent combination of anti-DHPPA-BSA and DHPAA-AP in a direct ELISA (IC50=1.5μmol/L), and for DHBA using a reagent combination of anti-DHBA-OV and DHBA-AP (IC50=1.3μmol/L). Calibration was conducted in the linear range (0.3-27.4μmol/L), with limit of detection (LOD) 0.1μmol/L. Intra and inter CVs was in the range of 0.7-7.2% and 5.1-11.5%, respectively, for DHPPA and 1.3-9.4% and 3.5-20%, respectively, for DHBA. Mean recovery was 104% for DHPPA and 102% for DHBA. The ELISA method developed was then used for analysis of 120 urine samples from free-living men and women that had previously been analysed by gas chromatography-mass spectrometry (GC-MS). ELISA produced several-fold higher values than GC-MS. Application of high-resolution Orbitrap mass spectrometry (HR Orbitrap MS) allowed several compounds, including novel putative AR metabolites, to be identified, synthesised and confirmed as compounds with high ELISA cross-reactivity.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Supelco
Syringic acid, analytical standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard