Merck
Lysobacter agri sp. nov., a bacterium isolated from soil.
Antonie van Leeuwenhoek (2015-06-27)
Hina Singh, KyungHwa Won, Juan Du, Jung-Eun Yang, Shahina Akter, Ki-Young Kim, Tae-Hoo Yi
ABSTRACT
A bacterial strain, designated as THG-SKA3(T), was isolated from field soil of Kyung Hee University, South Korea. Cells of the isolate were observed to be Gram-negative, aerobic, rod-shaped and motile by gliding. The strain was found to grow optimally at 28 °C, at pH 7 and in absence of NaCl. Based on 16S rRNA gene sequence comparisons, strain THG-SKA3(T) shared highest sequence similarity with Lysobacter niastensis KACC 11588(T) followed by Lysobacter panacisoli KACC 17502(T), Lysobacter enzymogenes LMG 8762(T) and Lysobacter oryzae KCTC 22249(T). The G+C content of THG-SKA3(T) was determined to be 68.9 mol%. The DNA-DNA relatedness values between strain THG-SKA3(T) and its closest phylogenetic neighbors were below 25.0 %.The major polar lipids of strain THG-SKA3(T) were determined to be diphosphatidylglycerol, phosphatidylglycerol and phosphatidylethanolamine. The predominant respiratory quinone was identified as ubiquinone 8 (Q-8). The major cellular fatty acids were identified as branched chain iso-C15:0, iso-C16:0 and unsaturated iso-C17:1 ω9c. On the basis of polyphasic data presented, it is evident that strain THG-SKA3(T) represents a novel species of the genus Lysobacter, for which the name Lysobacter agri sp. nov. (type strain THG-SKA3(T) = KACC 18283(T) = CSCTCC AB 2015126(T)) is proposed.
Product Description
SAFC
Ferric citrate
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SAFC
L-Tyrosine
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O