Merck
Are biological agents toxic to human chondrocytes and osteocytes?
Journal of orthopaedic surgery and research (2015-08-01)
Mehmet Isyar, Bulent Bilir, Ibrahim Yilmaz, Selami Cakmak, Duygu Yasar Sirin, Aliye Yildirim Guzelant, Mahir Mahirogullari
ABSTRACT
The aim of the present study is to investigate the effects of biological agents (BAs) on human chondrocytes and osteocytes in vitro. Primary cell cultures obtained from gonarthrosis patients were divided into four groups, two of which were designated as control cultures of chondrocyte and osteocyte, and the other two groups were exposed to BAs administered via the culture medium. Cultured cells were characterized by immunophenotyping. Before and after administration of the agents, the cultures were observed by inverted and environmental scanning electron microscopy (ESEM). The number of live cells and the proliferation rate were monitored by MTT assay. Rituximab and adalimumab were the least toxic agents to chondrocytes, whereas adalimumab and etanercept were to osteocytes. During periods of intense active inflammation, the concentration of the preferred BAs after inhibition of inflammation needs to be emphasized when their effects on cartilage and bone tissue are considered at the cellular level if the clinical practice is to continue.
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis