Merck
  • Bifidobacteria inhibit the inflammatory response induced by gliadins in intestinal epithelial cells via modifications of toxic peptide generation during digestion.
Bifidobacteria inhibit the inflammatory response induced by gliadins in intestinal epithelial cells via modifications of toxic peptide generation during digestion.
Journal of cellular biochemistry (2010-01-07)
J M Laparra, Y Sanz
ABSTRACT
Celiac disease (CD) is a chronic enteropathy triggered by intake of gliadin, the toxic component of gluten. This study aims at evaluating the capacity of different Bifidobacterium strains to counteract the inflammatory effects of gliadin-derived peptides in intestinal epithelial (Caco-2) cells. A commercial extract of several gliadin (Gld) types (alpha, beta, gamma, [symbol: see text] ) was subjected to in vitro gastrointestinal digestion (pepsin at pH 3, pancreatin-bile at pH 6), inoculated or not with cell suspensions (10(8) colony forming units/ml) of either B. animalis IATA-A2, B. longum IATA-ES1, or B. bifidum IATA-ES2, in a bicameral system. The generated gliadin-derived peptides were identified by reverse phase-HPLC-ESI-MS/MS. Caco-2 cell cultures were exposed to the different gliadin peptide digestions (0.25 mg protein/ml), and the mRNA expression of nuclear factor kappa-B (NF-kappaB), tumor necrosis factor alpha (TNF-alpha), and chemokine CXCR3 receptor were analyzed by semi-quantitative reverse transcriptase-polymerase chain reaction (RT-PCR) in stimulated cells. The production of the pro-inflammatory markers NF-kappaB p50, TNF-alpha, and IL-1beta (interleukine 1beta) by Caco-2 cells was also determined by ELISA. The peptides from gliadin digestions inoculated with bifidobacteria did not exhibit the toxic amino acid sequences identified in those noninoculated (alpha/beta-Gld [158-164] and alpha/beta-Gld [122-141]). The RT-PCR analysis evidenced a down-regulation in mRNA expression of pro-inflammatory biomarkers. Consistent with these results the production of NF-kappaB, TNF-alpha, and IL-1beta was reduced (18.2-22.4%, 28.0-64.8%, and abolished, respectively) in cell cultures exposed to gliadin digestions inoculated with bifidobacteria. Therefore, bifidobacteria change the gliadin-derived peptide pattern and, thereby, attenuate their pro-inflammatory effects on Caco-2 cells.
Sigma-Aldrich
Gliadin from wheat
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein