Merck
  • Uptake of glucose-conjugated MGMT inhibitors in cancer cells: role of flippases and type IV P-type ATPases.
Uptake of glucose-conjugated MGMT inhibitors in cancer cells: role of flippases and type IV P-type ATPases.
Scientific reports (2017-10-27)
Karl-Heinz Tomaszowski, Nadja Hellmann, Viviane Ponath, Hiroyuki Takatsu, Hye-Won Shin, Bernd Kaina
ABSTRACT
The DNA repair protein O
Sigma-Aldrich
O6-Benzylguanine, ≥98% (TLC), solid
Sigma-Aldrich
Lomeguatrib, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TMEM30A