Merck
GC Analysis of Mineral Spirits on TCEP, Fast GC Analysis
GC Analysis of Mineral Spirits on TCEP, Fast GC Analysis suitable for GC
CONDITIONS
carrier gas
hydrogen, 43 cm/sec
column
TCEP, 15 m x 0.10 mm I.D., 0.18 μm (28348-U)
detector
FID, 220 °C
inj. temp.
220 °C
injection
0.04 μL, 200:1 split
liner
4 mm I.D., split, cup design
oven
100 °C
sample
Low odor mineral spirits (from a local hardware store)