Accéder au contenu
Merck

ABN2252

Sigma-Aldrich

Anti-Neuroligin-4

Synonyme(s) :

Neuroligin Y

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme

Sélectionner une taille de conditionnement


A propos de cet article

Code UNSPSC :
12352203
Nomenclature NACRES :
NA.41
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider

Source biologique

rabbit

Niveau de qualité

Conjugué

unconjugated

Forme d'anticorps

purified antibody

Type de produit anticorps

primary antibodies

Clone

polyclonal

Espèces réactives

human

Conditionnement

antibody small pack of 100

Technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

Isotype

IgG

Séquence de l'épitope

C-terminal cytoplasmic domain

Numéro d'accès Protein ID

Numéro d'accès UniProt

Modification post-traductionnelle de la cible

unmodified

Informations sur le gène

human ... NLGN4Y(22829)

Description générale

Neuroligin-4 (UniProt: Q8NFZ3; also known as Neuroligin Y) is encoded by the NLGN4Y (also known as KIAA0951) gene (Gene ID: 22829) in human. Neuroligins are postsynaptic cell adhesion molecules that have crucial roles in synapse maturation and function. In humans, NLGN4X is located on the X chromosome and the NLGN4Y is localized on the Y chromosome and together they form an X-Y pair. They share about 97% amino acid identity; however, they are differentially regulated. Although several mutations in NLGN4X have been linked to autism, only one mutation has been identified in NLGN4Y gene. Despite their high degree of sequence identity, NLGN4Y exhibits severe deficit in trafficking to cell surface and related synaptic effects. This has been attributed to serine 93 in NLGN4Y. A substitution of serine to proline at residue 93 is shown to rescue it and increase its surface trafficking. Neuroligin-4 is a single-pass type I, homodimeric, membrane glycoprotein that is synthesized with a signal peptide (aa 1-43), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 44-676), a transmembrane domain (aa 677-697), and a cytoplasmic domain (aa 698-816). Its expression is observed in fetal and adult brain, prostate, and testis. It serves as a neuronal cell surface protein that is involved in cell-cell interactions. (Ref.: Nguyen, TA., et al. (2020). Neuron 106(5); 759-768; Marro, SG., et al. (2019). Neuron. 103(4); 617-626).

Immunogène

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal, cytoplasmic domain of human Neuroligin-4.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293T cells overexpressing Human Neuroligin-4.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Neuroligin-4 in lysate from HEK293T cells overexpressing Human Neuroligin-4.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Neuroligin-4 in Immunofluorescence applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Western Blotting Analysis: A representative lot detected Neuroligin-4 in Western Blotting applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Neuroligin-4 in Immunoprecipitation applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Actions biochimiques/physiologiques

This rabbit polyclonal antibody detects Neuroligin-4. It targets an epitope within 16 amino acids from the C-terminal, cytoplasmic domain.

Forme physique

Purified rabbit polyclonal antibody in buffer containing PBS, pH 7.4, without azide.

Notes préparatoires

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Autres remarques

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Clause de non-responsabilité

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Vous ne trouvez pas le bon produit ?  

Essayez notre Outil de sélection de produits.

Code de la classe de stockage

12 - Non Combustible Liquids

Classe de danger pour l'eau (WGK)

WGK 2

Point d'éclair (°F)

Not applicable

Point d'éclair (°C)

Not applicable


Certificats d'analyse (COA)

Recherchez un Certificats d'analyse (COA) en saisissant le numéro de lot du produit. Les numéros de lot figurent sur l'étiquette du produit après les mots "Lot" ou "Batch".

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique