Merck
All Photos(1)

GF08295532

Aluminum-magnesium alloy, Al97Mg3

foil, 6mm disks, 0.015mm thickness, as rolled

Synonym(s):
AL010225
NACRES:
NA.23

form

foil (disks - as rolled)

manufacturer/tradename

Goodfellow 082-955-32

size × thickness

6 mm × 0.015 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Packaging

10, 20, 50 ea

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service