PE1171
ampule of 2 × 2 mL
in acetone
environmental
matrix material
2-8°C
Danger
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Central nervous system
3 - Flammable liquids
WGK 2
1.4 °F - closed cup - Solvent
-17 °C - closed cup - Solvent
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service