Merck
All Photos(1)

SPE003

Supelco

BNAs in Soil - PT

Proficiency Testing Material

NACRES:
NA.24

Quality Level

packaging

pkg of 4 × 10 g

matrix

soil

application(s)

environmental

format

matrix material

General description

Bridged Nucleic Acids (BNAs) in Soil - PT proficiency testing material is produced in accordance with ISO/IEC 17043:2010. This product is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. We are the world leader in the manufacturing of real-world soil and sediment PT products.

Type of PT Scheme: Solids/Soils Hazardous Waste

Matrix: Soil

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of BNAs in Soil - PT can be found in the reporting packet which will be available to download at PT Portal.

Application

BNAs in Soil - PT is a soil/hazardous waste (soil/HW BNAs) proficiency testing material, suitable for testing the BNAs in soil samples.

Our soil PT samples behave as real samples; and, unless there is no alternative, are not simply spikes added to a blank soil. They are designed to meet the requirements of national, regional and state-specific accreditation bodies (TNI, USEPA, UKAS) for quality assurance and continuous improvement.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This sample consists of 10 g of soil containing base/neutrals and acids in soil.
  • Four samples have been provided for your convenience (multiple methods, multiple analysts, etc.).
  • The soil has been chemically stabilized with 1 mL of acetone to minimize degradation of the sample.
  • The evaluation of the results will be based on study statistics as required by TNI.
  • The standard may not contain all the compounds listed on the Report Form in accordance with TNI standards.
  • For analytes not detected, report as zero “0” or a “<” and your detection limit or “<” PTRL.


Sample Preparation

  • Extract the complete contents of a single vial. Transfer entire contents of one vial to extraction vessel. Rinse vial and cap with extraction solvent.
  • Note: Sample extracts and calibration solutions should be in the same solvent.
  • Analyze the Extract using the appropriate Semi-Volatile Organics method for which you are seeking accreditation.
  • Determine the concentration of the analytes listed and report your results on the organic analysis data sheet provided.
  • The method used must be reported.
  • Report all results on a wet weight basis, do not correct for moisture.
  • Assume a 10 g sample size for all calculations.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)

Soils
LPTP22-S1 January 26 - March 11, 2022
LPTP22-S2 April 6 - May 20, 2022
LPTP22-S3 July 20 - September 4, 2022
LPTP22-S4 October 5 - November 18, 2022

LPTP23-S2 April 5 – May 19, 2023
LPTP23-S4 October 4 – November 17, 2023


Quick-Turn Study (on demand proficiency testthat is offered depending on lot availability)

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene 1000-12000 μg/kg
    Acenaphthylene 1000-12000 μg/kg
    4-Chloro-3-methylphenol 1000-12000 μg/kg
    Acetophenone 500-15000 μg/kg
    2-Amino-1methylbenzene 500-15000 μg/kg
    Aniline 1500-15000 μg/kg
    Anthracene 1000-12000 μg/kg
    Atrazine 500-15000 μg/kg
    Benzidine 1500-15000 μg/kg
    Benzo(a)anthracene 1000-12000 μg/kg
    Benzo[b]fluoranthene 1000-12000 μg/kg
    Benzo[k]fluoranthene 1000-12000 μg/kg
    Benzo[ghi]perylene 1000-12000 μg/kg
    Benzo[a]pyrene 1000-12000 μg/kg
    Benzoic acid 1500-15000 μg/kg
    Benzyl alcohol 1500-15000 μg/kg
    Biphenyl 500-15000 μg/kg
    4-Bromodiphenyl ether 1500-15000 μg/kg
    Butyl benzyl phthalate 1000-12000 μg/kg
    Carbazole 1500-15000 μg/kg
    4-Chloro-3-methylphenol 1500-15000 μg/kg
    4-Chloroaniline 1500-15000 μg/kg
    Bis(2-chloroethoxy)methane 1000-12000 μg/kg
    Bis(2-chloroethyl) ether 1500-15000 μg/kg
    Bis-(2-chloroisopropyl) ether 1500-15000 μg/kg
    Caprolactam 500-15000 μg/kg
    1-Chloronaphthalene 1500-15000 μg/kg
    2-Chloronaphthalene 1000-12000 μg/kg
    2-Chlorophenol 1500-15000 μg/kg
    4-Chlorodiphenyl ether 1000-12000 μg/kg
    Chrysene 1000-12000 μg/kg
    Dibenzo(a,h)anthracene 1000-12000 μg/kg
    Dibenzofuran 1500-15000 μg/kg
    Di-n-butyl phthalate 1000-12000 μg/kg
    1,2-Dichlorobenzene 1500-15000 μg/kg
    1,3-Dichlorobenzene 1500-15000 μg/kg
    1,4-Dichlorobenzene 1500-15000 μg/kg
    3,3′-Dichlorobenzidine 1500-15000 μg/kg
    2,4-Dichlorophenol 1500-15000 μg/kg
    2,6-Dichlorophenol 1500-15000 μg/kg
    Diethyl phthalate 1000-12000 μg/kg
    2,3-Dimethylphenol 1000-20000 μg/kg
    2,4-Dimethylphenol 3000-15000 μg/kg
    2,5-Dimethylphenol 1000-20000 μg/kg
    2,6-Dimethylphenol 1000-20000 μg/kg
    3,4-Dimethylphenol 1000-20000 μg/kg
    3,5-Dimethylphenol 1000-20000 μg/kg
    Dimethyl phthalate 1000-12000 μg/kg
    2,4-Dinitrophenol 3000-15000 μg/kg
    2,4-Dinitrotoluene (2,4-DNT) 1500-15000 μg/kg
    2,6-Dinitrotoluene (2,6-DNT) 1500-15000 μg/kg
    Di-n-octyl phthalate 1000-12000 μg/kg
    bis(2-Ethylhexyl) phthalate (DEHP) 1500-15000 μg/kg
    Fluoranthene 1000-12000 μg/kg
    Fluorene 1000-12000 μg/kg
    Hexachlorobenzene 1500-15000 μg/kg
    Hexachlorobutadiene 1500-15000 μg/kg
    Hexachloroethane 1500-15000 μg/kg
    Hexachlorocyclopentadiene 1500-15000 μg/kg
    Indeno[1,2,3-c,d]pyrene solution 1000-12000 μg/kg
    Isophorone 1500-15000 μg/kg
    Maleic anhydride 1500-15000 μg/kg
    2-Methyl-4,6-dinitrophenol 3000-15000 μg/kg
    2-Methylnaphthalene (β) 1000-12000 μg/kg
    2-Methylphenol (o-Cresol) 3000-15000 μg/kg
    3-Methylphenol (m-Cresol) 3000-15000 μg/kg
    4-Methylphenol (p-Cresol) 3000-15000 μg/kg
    3+4-Methylphenol (m+p-Cresol) 500-15000 μg/kg
    Naphthalene 1000-12000 μg/kg
    Nitrobenzene 1500-15000 μg/kg
    2-Nitroaniline 1500-15000 μg/kg
    3-Nitroaniline 1500-15000 μg/kg
    4-Nitroaniline 1500-15000 μg/kg
    2-Nitrophenol 3000-15000 μg/kg
    3-Nitrophenol 1500-15000 μg/kg
    4-Nitrophenol 3000-15000 μg/kg
    N-Nitrosodimethylamine 1500-15000 μg/kg
    N-Nitrosodiphenylamine 1500-15000 μg/kg
    n-Nitroso-di-n-propylamine 1500-15000 μg/kg
    Pentachlorobenzene 1500-15000 μg/kg
    Pentachlorophenol 3000-15000 μg/kg
    Phenanthrene 1000-12000 μg/kg
    Phenol 1500-15000 μg/kg
    Pyrene 1000-12000 μg/kg
    1,2,3,4-Tetrachlorobenzene 1500-15000 μg/kg
    1,2,3,5-Tetrachlorobenzene 1500-15000 μg/kg
    1,2,4,5-Tetrachlorobenzene 1500-15000 μg/kg
    2,3,4,6-Tetrachlorophenol 1500-15000 μg/kg
    1,2,3-Trichlorobenzene 1500-15000 μg/kg
    1,2,4-Trichlorobenzene 1500-15000 μg/kg
    1,3,5-Trichlorobenzene 1500-15000 μg/kg
    2,4,5-Trichlorophenol 1500-15000 μg/kg
    2,4,6-Trichlorophenol 1500-15000 μg/kg
    Benzaldehyde 500-15000 μg/kg
    See All (94)

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service