Merck
All Photos(1)

Documents

P7957

Sigma-Aldrich

C-24 Sequencing Primer

Sign Into View Organizational & Contract Pricing

MDL number:
NACRES:
NA.85

grade

for molecular biology

Quality Level

form

solid

shipped in

dry ice

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P5350P1243908282
Sigma-Aldrich

Sigma-Aldrich

P7957

C-24 Sequencing Primer

form

solid

form

-

form

-

form

solid

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

C-24 Sequencing Primer is a chemically synthesized, 24 base, single strand oligodeoxyribonucleotide designed for dsDNA or ssDNA sequencing of FLAG® fusion proteins at the C-terminus of E. coli pFLAG expression vectors.

Application

Sequencing primers are used for double-stranded DNA sequencing of inserts in bacterial FLAG expression vectors.
The C-24 sequencing primer is suitable for sequencing from the C-terminus of E. coli pFLAG expression vectors.

Components

The primer is supplied at an initial concentration of 5 uM in 1xTE buffer. The nucleotide sequence is as follows: 5′
′-OH-CTG-TAT-CAG-GCT-GAA-AAT-CTT-CTC-3′−ΟΗ

Other Notes

(5′-CTG TAT CAG GCT GAA AAT CTT CTC-3′)

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service