All Photos(1)
22281
Valco® Precision Sampling Syringe, Series C, Removable needle
volume 2000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
material
bevel tip stainless steel needle (point style 2)
description
Series C
feature
removable needle Series C
packaging
pkg of 1 ea
needle L × O.D. × I.D.
2 in. × 0.029 in. × 0.012 in.
volume
2000 μL
Related Categories
General description
Series C - Removable Needle: A heavy duty gas syringe, it features a positive rear plunger stop and a special low-dead
volume, removable needle. No sample storage capacity.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Certificate of Analysis
Certificate of Origin