All Photos(1)
27316
Supelco
Vial, Chromacol
PTFE (non-disposable sleeve), O.D. × H 12 mm × 32 mm, for use with Supelco part numbers 27312, 27313, 27314, 27315, pkg of 40 ea
material
PTFE (non-disposable sleeve)
feature
closure type crimp top vial
packaging
pkg of 40 ea
O.D. × H
12 mm × 32 mm
fitting
thread for 8 mm
compatibility
for use with Supelco part numbers 27312, 27313, 27314, 27315
Related Categories
Certificate of Analysis
Certificate of Origin
Jenna Shiels et al.
PLoS neglected tropical diseases, 14(7), e0008470-e0008470 (2020-07-10)
Sm16, also known as SPO-1 and SmSLP, is a low molecular weight protein (~16kDa) secreted by the digenean trematode parasite Schistosoma mansoni, one of the main causative agents of human schistosomiasis. The molecule is secreted from the acetabular gland of
Simon R Carlile et al.
Scientific reports, 9(1), 6071-6071 (2019-04-17)
We investigated the anti-inflammatory and antibacterial activities of Hc-cath, a cathelicidin peptide derived from the venom of the sea snake, Hydrophis cyanocyntus, using in vivo models of inflammation and infection. Hc-cath function was evaluated in in vitro, in vivo in
Jlenia Brunetti et al.
The Journal of biological chemistry, 291(49), 25742-25748 (2016-10-21)
The synthetic antimicrobial peptide SET-M33 has strong activity against bacterial infections caused by Gram-negative bacteria. It is currently in preclinical development as a new drug to treat lung infections caused by Gram-negative bacteria. Here we report its strong anti-inflammatory activity
Leila Quercini et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(1), 192-207 (2020-01-10)
The peptide sequence KKIRVRLSA was synthesized in a dimeric structure (SET-M33DIM) and evaluated as a candidate drug for infections due to multidrug-resistant (MDR) Gram-negative pathogens. SET-M33DIM showed significant antibacterial activity against MDR strains of Klebsiella pneumoniae, Acinetobacter baumannii, and Escherichia