Merck
All Photos(1)

47990-U

Supelco

EPA TCL Hazardous Substances Mix (12 cmpds)

certified reference material, 2000 μg/mL each component in methylene chloride

NACRES:
NA.24

Quality Level

grade

certified reference material
TraceCERT®

Agency

EPA
610

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

Certified reference material (CRM) standards such as EPA TCL Hazardous Substances Mix (12 cmpds) are specifically designed for monitoring organic chemicals on the US EPA′s Target Compound List (TCL) in water, sediment, or soil from hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Aniline
    Benzoic acid
    Benzyl alcohol
    4-Chloroaniline
    o-Cresol
    p-Cresol
    Dibenzofuran
    2-Methylnaphthalene
    2-Nitroaniline
    3-Nitroaniline
    4-Nitroaniline
    2,4,5-Trichlorophenol
    See All (12)

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Blood, Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service