Merck
All Photos(1)

861216

Supelco

EPA 8270 Base Neutral Calibration Mix 2

certified reference material, 2000 μg/mL each component in dichloromethane

Quality Level

grade

certified reference material
TraceCERT®

Agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Aniline
    Anthracene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Benzyl alcohol
    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chloroaniline
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    2-Chloronaphthalene
    Chrysene
    Dibenz[a,h]anthracene
    Dibenzofuran
    Dibutyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Diethyl phthalate
    N,N-Dimethylaniline
    Dimethyl phthalate
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Di-n-octyl phthalate
    1,4-Dioxane
    Diphenyl ether
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Hydrazobenzene
    Indeno[1,2,3-cd]pyrene
    Isophorone
    2-Methylnaphthalene
    Naphthalene
    2-Nitroaniline
    3-Nitroaniline
    4-Nitroaniline
    Nitrobenzene
    N-Nitrosodimethylamine
    N-Nitrosodi-n-propylamine
    Phenanthrene
    Pyrene
    Pyridine trifluoroacetate
    1,2,4-Trichlorobenzene
    See All (56)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Blood, Respiratory system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service