MX
EN
Applications
Products
Services
Support
Home
Search Results
Benzalkonium Chloride 10% Solution
(1)
Benzalkonium Chloride 10% Solution
Synonyms:
Alkylbenzyldimethylammonium chloride, Benzalkonium chloride solution
CAS Number:
63449-41-2
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
PHR1371
Benzalkonium Chloride 10% Solution, Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing