Products
MX
EN
Products
Applications
Products
Services
Support
Home
Search Results
Collagen from rat tail
(2)
Collagen from rat tail
CAS Number:
9007-34-5
EC Number:
232-697-4
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
C7661
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Expand
View Pricing
C8897
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Expand
View Pricing