Merck
COMU® 97%

COMU®

COMU® 97%
Synonyms:
(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholino-carbenium hexafluorophosphate, COMU
Empirical Formula (Hill Notation):
C12H19F6N4O4P
CAS Number:
Molecular Weight:
428.27

Products

Brand
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Reaction Type

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2