MX
EN
Applications
Products
Services
Support
Home
Search Results
Liquified Phenol
(1)
Liquified Phenol
Synonyms:
Phenol solution
Empirical Formula (Hill Notation):
C
6
H
6
O
CAS Number:
108-95-2
Molecular Weight:
94.11
EC Number:
203-632-7
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
P9346
Liquified Phenol, ≥89.0%
Expand
View Pricing