MX
EN
Applications
Products
Services
Support
Home
Search Results
Starch, soluble
(1)
Starch, soluble
Linear Formula:
(C
6
H
10
O
5
)
n
EC Number:
232-679-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
S9765
Starch, soluble, ACS reagent
Expand
View Pricing