Merck
Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem TRIS base is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem

Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem TRIS base is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.
Synonyms:
Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Tris Base, Molecular Biology Grade - CAS 77-86-1 - Calbiochem, TRIS base is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.