  • Search Results
  • The mammalian CHORD-containing protein melusin is a stress response protein interacting with Hsp90 and Sgt1.
The mammalian CHORD-containing protein melusin is a stress response protein interacting with Hsp90 and Sgt1.
FEBS letters (2008-05-14)
Mauro Sbroggiò, Roberta Ferretti, Elena Percivalle, Malgorzata Gutkowska, Alicja Zylicz, Wojciech Michowski, Jacek Kuznicki, Federica Accornero, Beniamina Pacchioni, Gerolamo Lanfranchi, Jorg Hamm, Emilia Turco, Lorenzo Silengo, Guido Tarone, Mara Brancaccio
ABSTRACT
Melusin is a mammalian muscle specific CHORD containing protein capable of activating signal transduction pathways leading to cardiomyocytes hypertrophy in response to mechanical stress. To define melusin function we searched for molecular partners possibly involved in melusin dependent signal transduction. Here we show that melusin and heat shock proteins are co-regulated. Moreover, melusin directly binds to Hsp90, a ubiquitous chaperone involved in regulating several signaling pathways. In addition, melusin interacts with Sgt1, an Hsp90 binding molecule. Melusin does not behave as an Hsp90 substrate but rather as a chaperone capable to protect citrate synthase from heat induced aggregation. These results describe melusin as a new component of the Hsp90 chaperone machinery.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)