  • Search Results
  • The Wnt-Driven Mll1 Epigenome Regulates Salivary Gland and Head and Neck Cancer.
The Wnt-Driven Mll1 Epigenome Regulates Salivary Gland and Head and Neck Cancer.
Cell reports (2019-01-10)
Qionghua Zhu, Liang Fang, Julian Heuberger, Andrea Kranz, Jörg Schipper, Kathrin Scheckenbach, Ramon Oliveira Vidal, Daniele Yumi Sunaga-Franze, Marion Müller, Annika Wulf-Goldenberg, Sascha Sauer, Walter Birchmeier
ABSTRACT
We identified a regulatory system that acts downstream of Wnt/β-catenin signaling in salivary gland and head and neck carcinomas. We show in a mouse tumor model of K14-Cre-induced Wnt/β-catenin gain-of-function and Bmpr1a loss-of-function mutations that tumor-propagating cells exhibit increased Mll1 activity and genome-wide increased H3K4 tri-methylation at promoters. Null mutations of Mll1 in tumor mice and in xenotransplanted human head and neck tumors resulted in loss of self-renewal of tumor-propagating cells and in block of tumor formation but did not alter normal tissue homeostasis. CRISPR/Cas9 mutagenesis and pharmacological interference of Mll1 at sequences that inhibit essential protein-protein interactions or the SET enzyme active site also blocked the self-renewal of mouse and human tumor-propagating cells. Our work provides strong genetic evidence for a crucial role of Mll1 in solid tumors. Moreover, inhibitors targeting specific Mll1 interactions might offer additional directions for therapies to treat these aggressive tumors.
Product Description
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder