  • Search Results
  • A novel type of organometallic 2-R-2,4-dihydro-1H-3,1-benzoxazine with R = [M(η5-C5H4)(CO)3] (M = Re or Mn) units. Experimental and computational studies of the effect of substituent R on ring-chain tautomerism.
A novel type of organometallic 2-R-2,4-dihydro-1H-3,1-benzoxazine with R = [M(η5-C5H4)(CO)3] (M = Re or Mn) units. Experimental and computational studies of the effect of substituent R on ring-chain tautomerism.
Dalton transactions (Cambridge, England : 2003) (2019-01-04)
Juan Oyarzo, Ramón Bosque, Patricia Toro, Carlos P Silva, Rodrigo Arancibia, Mercè Font-Bardía, Vania Artigas, Carme Calvis, Ramon Messeguer, A Hugo Klahn, Concepción López
ABSTRACT
The syntheses, characterization, X-ray crystal structures, electrochemical properties and anticancer and antichagasic activities of the first examples of 2-substituted 2,4-dihydro-1H-3,1-benzoxazines with half-sandwich organometallic arrays, [M(η5-C5H4)(CO)3] (M = Re or Mn), at position-2 are described. Experimental and computational studies based on DFT calculations on the open forms [Schiff bases of general formulae R-CH[double bond, length as m-dash]N-C6H4-2-CH2OH] (5), with R = ferrocenyl (a), phenyl (b), cyrhetrenyl (c) or cymantrenyl (d), and their tautomeric forms (2-substituted 2,4-dihydro-1H-3,1 benzoxazines) have allowed us to establish the influence of substituents a-d and solvents on: (a) the extent of tautomeric equilibria (5a-5d) ↔ (6a-6d) and (b) their electrochemical properties and the electronic distribution on the open and closed forms. Despite the formal similarity between 6c and 6d, their anticancer and antiparasitic activities are markedly different. Compound 6d is inactive in the HCT116, MDA-MB231 and MCF7 cancer cell lines, but 6c shows moderate activity in the latter cell line, while the Mn(i) complex (6d) is a more potent anti-Trypanosoma cruzi agent than its Re(i) analogue (6c).
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture