  • Search Results
  • Down-regulation of interleukin-33 expression in oligodendrocyte precursor cells impairs oligodendrocyte lineage progression.
Down-regulation of interleukin-33 expression in oligodendrocyte precursor cells impairs oligodendrocyte lineage progression.
Journal of neurochemistry (2019-06-06)
Hsin-Yu Sung, Wei-Yu Chen, Hui-Ting Huang, Chih-Yen Wang, Song-Bin Chang, Shun-Fen Tzeng
ABSTRACT
Interleukin-33 (IL-33), a member of the IL1 family, has been found to be expressed in oligodendrocytes (OLGs) and released as an alarmin from injured OLGs to work on other glial cell-types in the central nervous system. However, its functional role in OLGs remains unclear. Herein, we present that IL-33 was mainly expressed in the nucleus of CC1+ -oligodendrocytes (OLGs) in mouse and rat corpus callosum, as well as NG2+ -oligodendrocyte precursor cells (OPCs). The in vitro study indicated that the amount of IL-33 expressing in OPCs was higher when compared to that detected in OLGs. Results from the experiments using lentivirus-mediated shRNA delivery against IL-33 expression (IL33-KD) in OPCs showed that IL33-KD reduced the differentiation of OLGs into mature OLGs along with the down-regulation of OLG differentiation-related genes and mature OLG marker proteins, myelin basic protein (MBP) and proteolipid protein (PLP). Alternatively, we observed reduced differentiation of OLGs that were prepared from the brains of IL-33 gene knockout (IL33-KO) mice with anxiolytic-like behavior. Observations were correlated with the results showing lower levels of MBP and PLP in IL33-KO cultures than those detected in the control cultures prepared from wildtype (WT) mice. Transmission Electron Microscopy (TEM) analysis revealed that the myelin structures in the corpus callosum of the IL33-KO mice were impaired compared to those observed in the WT mice. Overall, this study provides important evidence that declined expression of IL-33 in OPCs suppresses the maturation of OLGs. Moreover, gene deficiency of IL-33 can disrupt OLG maturation and interfere with myelin compaction. Cover Image for this issue: doi: 10.1111/jnc.14522.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000
Sigma-Aldrich
apo-Transferrin human, ≥95% protein basis (biuret)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%