  • Search Results
  • The SINEB1 element in the long non-coding RNA Malat1 is necessary for TDP-43 proteostasis.
The SINEB1 element in the long non-coding RNA Malat1 is necessary for TDP-43 proteostasis.
Nucleic acids research (2019-12-22)
Tuan M Nguyen, Elena B Kabotyanski, Lucas C Reineke, Jiaofang Shao, Feng Xiong, Joo-Hyung Lee, Julien Dubrulle, Hannah Johnson, Fabio Stossi, Phoebe S Tsoi, Kyoung-Jae Choi, Alexander G Ellis, Na Zhao, Jin Cao, Oluwatoyosi Adewunmi, Josephine C Ferreon, Allan Chris M Ferreon, Joel R Neilson, Michael A Mancini, Xi Chen, Jongchan Kim, Li Ma, Wenbo Li, Jeffrey M Rosen
ABSTRACT
Transposable elements (TEs) comprise a large proportion of long non-coding RNAs (lncRNAs). Here, we employed CRISPR to delete a short interspersed nuclear element (SINE) in Malat1, a cancer-associated lncRNA, to investigate its significance in cellular physiology. We show that Malat1 with a SINE deletion forms diffuse nuclear speckles and is frequently translocated to the cytoplasm. SINE-deleted cells exhibit an activated unfolded protein response and PKR and markedly increased DNA damage and apoptosis caused by dysregulation of TDP-43 localization and formation of cytotoxic inclusions. TDP-43 binds stronger to Malat1 without the SINE and is likely 'hijacked' by cytoplasmic Malat1 to the cytoplasm, resulting in the depletion of nuclear TDP-43 and redistribution of TDP-43 binding to repetitive element transcripts and mRNAs encoding mitotic and nuclear-cytoplasmic regulators. The SINE promotes Malat1 nuclear retention by facilitating Malat1 binding to HNRNPK, a protein that drives RNA nuclear retention, potentially through direct interactions of the SINE with KHDRBS1 and TRA2A, which bind to HNRNPK. Losing these RNA-protein interactions due to the SINE deletion likely creates more available TDP-43 binding sites on Malat1 and subsequent TDP-43 aggregation. These results highlight the significance of lncRNA TEs in TDP-43 proteostasis with potential implications in both cancer and neurodegenerative diseases.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid