  • Search Results
  • Hepatoprotective amide constituents from the fruit of Piper chaba: Structural requirements, mode of action, and new amides.
Hepatoprotective amide constituents from the fruit of Piper chaba: Structural requirements, mode of action, and new amides.
Bioorganic & medicinal chemistry (2009-09-25)
Hisashi Matsuda, Kiyofumi Ninomiya, Toshio Morikawa, Daisuke Yasuda, Itadaki Yamaguchi, Masayuki Yoshikawa
ABSTRACT
The 80% aqueous acetone extract from the fruit of Piper chaba (Piperaceae) was found to have hepatoprotective effects on D-galactosamine (D-GalN)/lipopolysaccharide-induced liver injury in mice. From the ethyl acetate-soluble fraction, three new amides, piperchabamides E, G, and H, 33 amides, and four aromatic constituents were isolated. Among the isolates, several amide constituents inhibited D-GalN/tumor necrosis factor-alpha (TNF-alpha)-induced death of hepatocytes, and the following structural requirements were suggested: (i) the amide moiety is essential for potent activity; and (ii) the 1,9-decadiene structure between the benzene ring and the amide moiety tended to enhance the activity. Moreover, a principal constituent, piperine, exhibited strong in vivo hepatoprotective effects at doses of 5 and 10 mg/kg, po and its mode of action was suggested to depend on the reduced sensitivity of hepatocytes to TNF-alpha.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Piperine, ≥97%
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Piperine, ≥95%, FG
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Hydrocortisone, VETRANAL®, analytical standard