  • Search Results
  • Development of an in silico model for human skin permeation based on a Franz cell skin permeability assay.
Development of an in silico model for human skin permeation based on a Franz cell skin permeability assay.
Bioorganic & medicinal chemistry letters (2009-12-08)
Pil H Lee, Robert Conradi, Veerabahu Shanmugasundaram
ABSTRACT
A multiple linear regression QSAR model was developed based on a set of 61 compounds with internally consistent permeability data measured across Franz cell. The data was normalized using a mean permeability value of a reference compound, 3-isobutyl-1-methylxanthine (IBMX). The QSAR model contained only five simple descriptors and had a correlation coefficient, r(2) of 0.77 between experimental and calculated values for skin permeability. The mean absolute error (MAE) was 0.3 for the entire set and the cross validation coefficient, q(2) was 0.71. The in silico skin permeability model was used as a filter for virtual libraries and to optimize skin permeation of specific compounds for several dermatology discovery projects.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Corticosterone, ≥92%
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Clonidine hydrochloride, solid