  • Search Results
  • GSH-Independent Induction of ER Stress during Hypoglycaemia in the Retinal Cells of Mice.
GSH-Independent Induction of ER Stress during Hypoglycaemia in the Retinal Cells of Mice.
Journal of clinical medicine (2021-07-03)
Daria Fresia, Enrica Cannizzaro, Angelica Borgo, Raphaël Roduit
ABSTRACT
Glucose is one of the most important metabolic substrates of the retina, and glycaemic imbalances can lead to serious side effects, including retinopathy. We previously showed that hypoglycaemia induces retinal cell death in mice, as well as the implication of glutathione (GSH) in this process. This study aimed to analyse the role of low glucose-induced decrease in GSH levels in endoplasmic reticulum (ER) stress. We cultured 661W photoreceptor-like cells under various glucose conditions and analysed ER stress markers at the mRNA and protein levels. We used the ERAI ("ER stress-activated indicator") mouse model to test ER stress in both ex vivo, on retinal explants, or in vivo, in mice subjected to hypoglycaemia. Moreover, we used buthionine sulfoximine (BSO) and glutamate cysteine ligase (Gclm)-KO mice as models of low GSH to test its effects on ER stress. We show that the unfolded protein response (UPR) is triggered in 661W cells and in ERAI mice under hypoglycaemic conditions. Low GSH levels promote cell death, but have no impact on ER stress. We concluded that low glucose levels induce ER stress independently of GSH levels. Inhibition of ER stress could prevent neurodegeneration, which seems to be an early event in the pathogenesis of diabetic retinopathy.
Sigma-Aldrich
Tunicamycin Ready Made Solution, 5 mg/mL in DMSO, from Streptomyces sp.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Monoclonal Anti-p130 antibody produced in mouse, clone KAB40, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
meta-Phosphoric acid, ACS reagent, chips, 33.5-36.5%