  • Search Results
  • Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy (2021-05-15)
Gülen Güney-Esken, Özgür Doğuş Erol, Burcu Pervin, Gülben Gürhan Sevinç, Tamer Önder, Elif Bilgiç, Petek Korkusuz, Ayşen Günel-Özcan, Duygu Uçkan-Çetinkaya, Fatima Aerts-Kaya
ABSTRACT
Griscelli syndrome type 2 (GS-2) is a rare, autosomal recessive immune deficiency syndrome caused by a mutation in the RAB27A gene, which results in the absence of a protein involved in vesicle trafficking and consequent loss of function of in particular cytotoxic T and NK cells. Induced pluripotent stem cells (iPSC) express genes associated with pluripotency, have the capacity for infinite expansion, and can differentiate into cells from all three germ layers. They can be induced using integrative or non-integrative systems for transfer of the Oct4, Sox2, Klf4, and cMyc (OSKM) transcription factors. To better understand the pathophysiology of GS-2 and to test novel treatment options, there is a need for an in vitro model of GS-2. Here, we generated iPSCs from 3 different GS-2 patients using lentiviral vectors. The iPSCs were characterized using flow cytometry and RT-PCR and tested for the expression of pluripotency markers. In vivo differentiation to cells from all three germlines was tested using a teratoma assay. In vitro differentiation of GS-2 iPSCs into hematopoietic stem and progenitor cells was done using Op9 feeder layers and specified media. All GS-2 iPSC clones displayed a normal karyotype (46XX or 46XY) and were shown to express the same RAB27A gene mutation that was present in the original somatic donor cells. GS-2 iPSCs expressed SSEA1, SSEA4, TRA-1-60, TRA-1-81, and OCT4 proteins, and SOX2, NANOG, and OCT4 expression were confirmed by RT-PCR. Differentiation capacity into cells from all three germ layers was confirmed using the teratoma assay. GS-2 iPSCs showed the capacity to differentiate into cells of the hematopoietic lineage. Using the lentiviral transfer of OSKM, we were able to generate different iPSC clones from 3 GS-2 patients. These cells can be used in future studies for the development of novel treatment options and to study the pathophysiology of GS-2 disease.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MCDB 201 Medium, With trace elements, L-glutamine and 30 mM HEPES, powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)