  • Search Results
  • RNF111/Arkadia is a SUMO-targeted ubiquitin ligase that facilitates the DNA damage response.
RNF111/Arkadia is a SUMO-targeted ubiquitin ligase that facilitates the DNA damage response.
The Journal of cell biology (2013-06-12)
Sara L Poulsen, Rebecca K Hansen, Sebastian A Wagner, Loes van Cuijk, Gijsbert J van Belle, Werner Streicher, Mats Wikström, Chunaram Choudhary, Adriaan B Houtsmuller, Jurgen A Marteijn, Simon Bekker-Jensen, Niels Mailand
ABSTRACT
Protein modifications by ubiquitin and small ubiquitin-like modifier (SUMO) play key roles in cellular signaling pathways. SUMO-targeted ubiquitin ligases (STUbLs) directly couple these modifications by selectively recognizing SUMOylated target proteins through SUMO-interacting motifs (SIMs), promoting their K48-linked ubiquitylation and degradation. Only a single mammalian STUbL, RNF4, has been identified. We show that human RNF111/Arkadia is a new STUbL, which used three adjacent SIMs for specific recognition of poly-SUMO2/3 chains, and used Ubc13-Mms2 as a cognate E2 enzyme to promote nonproteolytic, K63-linked ubiquitylation of SUMOylated target proteins. We demonstrate that RNF111 promoted ubiquitylation of SUMOylated XPC (xeroderma pigmentosum C) protein, a central DNA damage recognition factor in nucleotide excision repair (NER) extensively regulated by ultraviolet (UV)-induced SUMOylation and ubiquitylation. Moreover, we show that RNF111 facilitated NER by regulating the recruitment of XPC to UV-damaged DNA. Our findings establish RNF111 as a new STUbL that directly links nonproteolytic ubiquitylation and SUMOylation in the DNA damage response.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Hydroquinidine, 95%
Sigma-Aldrich
Selenic acid solution, 40 wt. % in H2O, 99.95% trace metals basis