  • Search Results
  • Behavior training reverses asymmetry in hippocampal transcriptome of the cav3.2 knockout mice.
Behavior training reverses asymmetry in hippocampal transcriptome of the cav3.2 knockout mice.
PloS one (2015-03-15)
Ni-Chun Chung, Ying-Hsueh Huang, Chuan-Hsiung Chang, James C Liao, Chih-Hsien Yang, Chien-Chang Chen, Ingrid Y Liu
ABSTRACT
Homozygous Cav3.2 knockout mice, which are defective in the pore-forming subunit of a low voltage activated T-type calcium channel, have been documented to show impaired maintenance of late-phase long-term potentiation (L-LTP) and defective retrieval of context-associated fear memory. To investigate the role of Cav3.2 in global gene expression, we performed a microarray transcriptome study on the hippocampi of the Cav3.2-/- mice and their wild-type littermates, either naïve (untrained) or trace fear conditioned. We found a significant left-right asymmetric effect on the hippocampal transcriptome caused by the Cav3.2 knockout. Between the naive Cav3.2-/- and the naive wild-type mice, 3522 differentially expressed genes (DEGs) were found in the left hippocampus, but only 4 DEGs were found in the right hippocampus. Remarkably, the effect of Cav3.2 knockout was partially reversed by trace fear conditioning. The number of DEGs in the left hippocampus was reduced to 6 in the Cav3.2 knockout mice after trace fear conditioning, compared with the wild-type naïve mice. To our knowledge, these results demonstrate for the first time the asymmetric effects of the Cav3.2 and its partial reversal by behavior training on the hippocampal transcriptome.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer