  • Search Results
  • Combined Application of Antibrowning, Heat Treatment and Modified-Atmosphere Packaging to Extend the Shelf Life of Fresh-Cut Lotus Root.
Combined Application of Antibrowning, Heat Treatment and Modified-Atmosphere Packaging to Extend the Shelf Life of Fresh-Cut Lotus Root.
Journal of food science (2015-04-30)
Jihye Son, Jeong-Eun Hyun, Jo-Won Lee, Sun-Young Lee, BoKyung Moon
ABSTRACT
This work aimed to determine the effects of different concentrations of antibrowning treatments (that is, distilled water [DW], 1% ascorbic acid [AA], 0.5% chamomile [CM], and 1% AA + 0.5% CM) and heat-treatment (55 °C for 45 s) combined with packaging under 4 different modified-atmosphere gas compositions (that is, air, vacuum, 100% CO2 , 50% CO2 /50% N2 ) on the quality and microbiological characteristics of fresh-cut lotus root. The quality characteristics (that is, color, weight loss, texture, pH, polyphenoloxidase activity, and total phenolic content) of the AA + CM-dipped sample in 100% CO2 packaging were maintained significantly better than those of the other samples (P < 0.05). The microbiological counts observed in the DW-dipped sample during storage were higher than those of the AA, CM, and AA + CM samples, and heat-treatment retarded the microbiological deterioration of fresh-cut lotus root. Therefore, the results revealed that dipping in an antibrowning treatment (AA + CM), and 100% CO2 MAP with heat treatment effectively extend the shelf life of fresh-cut lotus root to 21 d at 5 °C.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, suitable for plant cell culture, BioReagent, ≥99%, powder
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%