  • Search Results
  • Integrating Substrateless Electrospinning with Textile Technology for Creating Biodegradable Three-Dimensional Structures.
Integrating Substrateless Electrospinning with Textile Technology for Creating Biodegradable Three-Dimensional Structures.
Nano letters (2015-07-28)
John Joseph, Shantikumar V Nair, Deepthy Menon
ABSTRACT
The present study describes a unique way of integrating substrateless electrospinning process with textile technology. We developed a new collector design that provided a pressure-driven, localized cotton-wool structure in free space from which continuous high strength yarns were drawn. An advantage of this integration was that the textile could be drug/dye loaded and be developed into a core-sheath architecture with greater functionality. This method could produce potential nanotextiles for various biomedical applications.
Product Description
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Cardiogreen, polymethine dye