  • Search Results
  • miR-190a inhibits epithelial-mesenchymal transition of hepatoma cells via targeting the long non-coding RNA treRNA.
miR-190a inhibits epithelial-mesenchymal transition of hepatoma cells via targeting the long non-coding RNA treRNA.
FEBS letters (2015-11-27)
Xinyu Wang, Yanli Ren, Xinyu Yang, Xiangyu Xiong, Sichong Han, Yunxia Ge, Wenting Pan, Liqing Zhou, Qipeng Yuan, Ming Yang
ABSTRACT
treRNA is a long non-coding RNA (lncRNA) involved in cancer progression. In this study, we show that miR-190a can silence treRNA post-transcriptionally. Suppression of treRNA by miR-190a led to significant changes of mesenchymal-epithelial transition markers and impaired migration and invasion capability of hepatoma cells. TCGA data indicated that miR-190a exhibited lower expression in hepatoma tissues, especially from patients with vascular tumor invasion, compared to normal tissues. Our results reveal the involvement of miR-190a-treRNA axis in hepatoma progression and shed light on lncRNA-based cancer therapies for hepatoma patients at high risk of metastasis.
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture