  • Search Results
  • A conserved influenza A virus nucleoprotein code controls specific viral genome packaging.
A conserved influenza A virus nucleoprotein code controls specific viral genome packaging.
Nature communications (2016-09-22)
Étori Aguiar Moreira, Anna Weber, Hardin Bolte, Larissa Kolesnikova, Sebastian Giese, Seema Lakdawala, Martin Beer, Gert Zimmer, Adolfo García-Sastre, Martin Schwemmle, Mindaugas Juozapaitis
ABSTRACT
Packaging of the eight genomic RNA segments of influenza A viruses (IAV) into viral particles is coordinated by segment-specific packaging sequences. How the packaging signals regulate the specific incorporation of each RNA segment into virions and whether other viral or host factors are involved in this process is unknown. Here, we show that distinct amino acids of the viral nucleoprotein (NP) are required for packaging of specific RNA segments. This was determined by studying the NP of a bat influenza A-like virus, HL17NL10, in the context of a conventional IAV (SC35M). Replacement of conserved SC35M NP residues by those of HL17NL10 NP resulted in RNA packaging defective IAV. Surprisingly, substitution of these conserved SC35M amino acids with HL17NL10 NP residues led to IAV with altered packaging efficiencies for specific subsets of RNA segments. This suggests that NP harbours an amino acid code that dictates genome packaging into infectious virions.
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid