Merck
The RasGAP gene, RASAL2, is a tumor and metastasis suppressor.
Cancer cell (2013-09-14)
Sara Koenig McLaughlin, Sarah Naomi Olsen, Benjamin Dake, Thomas De Raedt, Elgene Lim, Roderick Terry Bronson, Rameen Beroukhim, Kornelia Polyak, Myles Brown, Charlotte Kuperwasser, Karen Cichowski
ABSTRACT
RAS genes are commonly mutated in cancer; however, RAS mutations are rare in breast cancer, despite frequent hyperactivation of Ras and ERK. Here, we report that the RasGAP gene, RASAL2, functions as a tumor and metastasis suppressor. RASAL2 is mutated or suppressed in human breast cancer, and RASAL2 ablation promotes tumor growth, progression, and metastasis in mouse models. In human breast cancer, RASAL2 loss is associated with metastatic disease; low RASAL2 levels correlate with recurrence of luminal B tumors; and RASAL2 ablation promotes metastasis of luminal mouse tumors. Additional data reveal a broader role for RASAL2 inactivation in other tumor types. These studies highlight the expanding role of RasGAPs and reveal an alternative mechanism of activating Ras in cancer.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution