  • Search Results
  • Mitochondrial dysfunction induces dendritic loss via eIF2α phosphorylation.
Mitochondrial dysfunction induces dendritic loss via eIF2α phosphorylation.
The Journal of cell biology (2017-02-18)
Taiichi Tsuyama, Asako Tsubouchi, Tadao Usui, Hiromi Imamura, Tadashi Uemura
ABSTRACT
Mitochondria are key contributors to the etiology of diseases associated with neuromuscular defects or neurodegeneration. How changes in cellular metabolism specifically impact neuronal intracellular processes and cause neuropathological events is still unclear. We here dissect the molecular mechanism by which mitochondrial dysfunction induced by Prel aberrant function mediates selective dendritic loss in
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine and potassium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture
Ouabain, European Pharmacopoeia (EP) Reference Standard